• USA Home
  • CDS004745 - (R)-4-N-Boc-piperazine-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
CDS004745 Sigma-Aldrich

(R)-4-N-Boc-piperazine-2-carboxylic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C10H18N2O4

  • Molecular Weight 230.26

  •  MDL number MFCD02179099

  •  PubChem Substance ID 329781443

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
SMILES string   CC(C)(C)OC(=O)N1CCN[C@H](C1)C(O)=O
InChI   1S/C10H18N2O4/c1-10(2,3)16-9(15)12-5-4-11-7(6-12)8(13)14/h7,11H,4-6H2,1-3H3,(H,13,14)/t7-/m1/s1
InChI key   YRYAXQJXMBETAT-SSDOTTSWSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?