• USA Home
  • CDS004961 - 2-((tetrahydrofuran-2-ylmethyl)amino)pyrimidine-5-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
CDS004961 Sigma-Aldrich

2-((tetrahydrofuran-2-ylmethyl)amino)pyrimidine-5-carbaldehyde

AldrichCPR

  • Empirical Formula (Hill Notation) C10H13N3O2

  • Molecular Weight 207.23

  •  MDL number MFCD09864455

  •  PubChem Substance ID 329781621

Purchase

Properties

Related Categories Building Blocks, C9 to C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
SMILES string   O=Cc1cnc(NCC2CCCO2)nc1
InChI   1S/C10H13N3O2/c14-7-8-4-11-10(12-5-8)13-6-9-2-1-3-15-9/h4-5,7,9H,1-3,6H2,(H,11,12,13)
InChI key   LRUPLPRMHCXMTM-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?