• USA Home
  • CDS005368 - 1-naphthyl trifluoromethyl ketone

EMAIL THIS PAGE TO A FRIEND
CDS005368 Sigma-Aldrich

1-naphthyl trifluoromethyl ketone

AldrichCPR

  • Empirical Formula (Hill Notation) C12H7F3O

  • Molecular Weight 224.18

  •  MDL number MFCD01320007

  •  PubChem Substance ID 329781892

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Chemical Synthesis, Fluorinated Building Blocks,
SMILES string   FC(F)(F)C(=O)c1cccc2ccccc12
InChI   1S/C12H7F3O/c13-12(14,15)11(16)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H
InChI key   VQCWOGKZDGCRES-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?