• USA Home
  • CDS006367 - 2,4-Dichloro-6-(4-fluorophenyl)pyrimidine

EMAIL THIS PAGE TO A FRIEND
CDS006367 Sigma-Aldrich

2,4-Dichloro-6-(4-fluorophenyl)pyrimidine

AldrichCPR

  • Empirical Formula (Hill Notation) C10H5Cl2FN2

  • Molecular Weight 243.06

  •  MDL number MFCD11043606

  •  PubChem Substance ID 329782757

Purchase

Properties

Related Categories Building Blocks, C9 to C11, Chemical Synthesis, Fluorinated Building Blocks, Halogenated Heterocycles,
SMILES string   Fc1ccc(cc1)-c2cc(Cl)nc(Cl)n2
InChI   1S/C10H5Cl2FN2/c11-9-5-8(14-10(12)15-9)6-1-3-7(13)4-2-6/h1-5H
InChI key   QQLFWYQWFPLRRS-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?