• USA Home
  • CDS006567 - 1-Isopropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline

EMAIL THIS PAGE TO A FRIEND
CDS006567 Sigma-Aldrich

1-Isopropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline

AldrichCPR

  • Empirical Formula (Hill Notation) C14H21NO2

  • Molecular Weight 235.32

  •  MDL number MFCD00231221

  •  PubChem Substance ID 329782926

Purchase

Properties

Related Categories Amines, Building Blocks, C14, Chemical Synthesis, Ethers,
SMILES string   COc1cc2CCNC(C(C)C)c2cc1OC
InChI   1S/C14H21NO2/c1-9(2)14-11-8-13(17-4)12(16-3)7-10(11)5-6-15-14/h7-9,14-15H,5-6H2,1-4H3
InChI key   IRWSLTSAQIJKQL-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?