• USA Home
  • CDS007908 - 2-sec-Butoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
CDS007908 Sigma-Aldrich

2-sec-Butoxybenzaldehyde

AldrichCPR

  • Empirical Formula (Hill Notation) C11H14O2

  • Molecular Weight 178.23

  •  MDL number MFCD02629688

  •  PubChem Substance ID 329783978

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Ethers, Organic Building Blocks,
SMILES string   CCC(C)Oc1ccccc1C=O
InChI   1S/C11H14O2/c1-3-9(2)13-11-7-5-4-6-10(11)8-12/h4-9H,3H2,1-2H3
InChI key   NHIGQSREBZKSAJ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?