• USA Home
  • CDS007966 - 5,6-dihydro-1,4-dioxine-2-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
CDS007966 Sigma-Aldrich

5,6-dihydro-1,4-dioxine-2-carbaldehyde

AldrichCPR

  • Empirical Formula (Hill Notation) C5H6O3

  • Molecular Weight 114.10

  •  MDL number MFCD00622394

  •  PubChem Substance ID 329783988

Purchase

Properties

Related Categories Building Blocks, C2 to C7, Chemical Synthesis, Ethers, Organic Building Blocks,
SMILES string   O=CC1=COCCO1
InChI   1S/C5H6O3/c6-3-5-4-7-1-2-8-5/h3-4H,1-2H2
InChI key   GXZRZOIXFFDVTD-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?