• USA Home
  • CDS008316 - (S)-n4-benzyl-2-(methylthioethyl)piperazine

EMAIL THIS PAGE TO A FRIEND
CDS008316 Sigma-Aldrich

(S)-n4-benzyl-2-(methylthioethyl)piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H22N2S

  • Molecular Weight 250.40

  •  MDL number MFCD03787924

  •  PubChem Substance ID 329784380

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
SMILES string   CSCC[C@H]1CN(CCN1)Cc2ccccc2
InChI   1S/C14H22N2S/c1-17-10-7-14-12-16(9-8-15-14)11-13-5-3-2-4-6-13/h2-6,14-15H,7-12H2,1H3/t14-/m0/s1
InChI key   KTJCLLHVCSLTGA-AWEZNQCLSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?