• USA Home
  • CDS008765 - 4-(2-methoxyphenoxy)benzoic acid

EMAIL THIS PAGE TO A FRIEND
CDS008765 Sigma-Aldrich

4-(2-methoxyphenoxy)benzoic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C14H12O4

  • Molecular Weight 244.24

  •  MDL number MFCD01570262

  •  PubChem Substance ID 329784771

Purchase

Properties

Related Categories Building Blocks, C14, Chemical Synthesis, Ethers, Organic Building Blocks,
SMILES string   COc1ccccc1Oc2ccc(cc2)C(O)=O
InChI   1S/C14H12O4/c1-17-12-4-2-3-5-13(12)18-11-8-6-10(7-9-11)14(15)16/h2-9H,1H3,(H,15,16)
InChI key   UYBHUZLSTOFTOG-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?