• USA Home
  • CDS008795 - [2-(4-methylpiperazinyl)-1-phenylethyl]methylamine

EMAIL THIS PAGE TO A FRIEND
CDS008795 Sigma-Aldrich

[2-(4-methylpiperazinyl)-1-phenylethyl]methylamine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H23N3

  • Molecular Weight 233.35

  •  MDL number MFCD01862535

  •  PubChem Substance ID 329784796

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
SMILES string   CNC(CN1CCN(C)CC1)c2ccccc2
InChI   1S/C14H23N3/c1-15-14(13-6-4-3-5-7-13)12-17-10-8-16(2)9-11-17/h3-7,14-15H,8-12H2,1-2H3
InChI key   KFLLDKIXAMGZLH-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?