• USA Home
  • CDS008879 - 2,4-dichloro-6-(3-chloro-4-fluorophenyl)pyrimidine

EMAIL THIS PAGE TO A FRIEND
CDS008879 Sigma-Aldrich

2,4-dichloro-6-(3-chloro-4-fluorophenyl)pyrimidine

AldrichCPR

  • Empirical Formula (Hill Notation) C10H4Cl3FN2

  • Molecular Weight 277.51

  •  MDL number MFCD11043668

  •  PubChem Substance ID 329784863

Purchase

Properties

Related Categories Building Blocks, C9 to C11, Chemical Synthesis, Fluorinated Building Blocks, Halogenated Heterocycles,
SMILES string   Fc1ccc(cc1Cl)-c2cc(Cl)nc(Cl)n2
InChI   1S/C10H4Cl3FN2/c11-6-3-5(1-2-7(6)14)8-4-9(12)16-10(13)15-8/h1-4H
InChI key   AVRMOBPNWJLZJM-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?