• USA Home
  • CDS011827 - 2-amino-4-methoxy-6-pyrrolidin-1-ylpyrimidine-5-carbaldehyde

EMAIL THIS PAGE TO A FRIEND
CDS011827 Sigma-Aldrich

2-amino-4-methoxy-6-pyrrolidin-1-ylpyrimidine-5-carbaldehyde

AldrichCPR

  • Empirical Formula (Hill Notation) C10H14N4O2

  • Molecular Weight 222.24

  •  MDL number MFCD09376440

  •  PubChem Substance ID 329787191

Purchase

Properties

Related Categories Building Blocks, C9 to C10, C9 to C11, Chemical Synthesis, Heterocyclic Building Blocks,
SMILES string   COc1nc(N)nc(N2CCCC2)c1C=O
InChI   1S/C10H14N4O2/c1-16-9-7(6-15)8(12-10(11)13-9)14-4-2-3-5-14/h6H,2-5H2,1H3,(H2,11,12,13)
InChI key   FRIOHBNVLBRJOL-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?