• USA Home
  • CDS012458 - 1-[2-(trifluoromethyl)quinol-4-yl]piperazine

EMAIL THIS PAGE TO A FRIEND
CDS012458 Sigma-Aldrich

1-[2-(trifluoromethyl)quinol-4-yl]piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H14F3N3

  • Molecular Weight 281.28

  •  MDL number MFCD00219765

  •  PubChem Substance ID 329787560

Purchase

Properties

Related Categories Building Blocks, C11 to C30, C14 to C15, Chemical Synthesis, Halogenated Heterocycles,
SMILES string   FC(F)(F)c1cc(N2CCNCC2)c3ccccc3n1
InChI   1S/C14H14F3N3/c15-14(16,17)13-9-12(20-7-5-18-6-8-20)10-3-1-2-4-11(10)19-13/h1-4,9,18H,5-8H2
InChI key   VCJGCFMRAFRJNE-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?