• USA Home
  • CDS012678 - n-butyl(4-(2-pyridyl)piperazinyl)formamide

EMAIL THIS PAGE TO A FRIEND
CDS012678 Sigma-Aldrich

n-butyl(4-(2-pyridyl)piperazinyl)formamide

AldrichCPR

  • Empirical Formula (Hill Notation) C14H22N4O

  • Molecular Weight 262.35

  •  MDL number MFCD01524911

  •  PubChem Substance ID 329787718

Purchase

Properties

Related Categories Building Blocks, C13 to C15, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks,
SMILES string   CCCCNC(=O)N1CCN(CC1)c2ccccn2
InChI   1S/C14H22N4O/c1-2-3-7-16-14(19)18-11-9-17(10-12-18)13-6-4-5-8-15-13/h4-6,8H,2-3,7,9-12H2,1H3,(H,16,19)
InChI key   NFTGOFKIERTOHA-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?