• USA Home
  • CDS014171 - 2-(2-pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol

EMAIL THIS PAGE TO A FRIEND
CDS014171 Sigma-Aldrich

2-(2-pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol

AldrichCPR

  • Empirical Formula (Hill Notation) C10H6F3N3O

  • Molecular Weight 241.17

  •  MDL number MFCD00139776

  •  PubChem Substance ID 329788753

Purchase

Properties

Related Categories Building Blocks, C10, C9 to C11, Chemical Synthesis, Heterocyclic Building Blocks,
SMILES string   Oc1cc(nc(n1)-c2ccccn2)C(F)(F)F
InChI   1S/C10H6F3N3O/c11-10(12,13)7-5-8(17)16-9(15-7)6-3-1-2-4-14-6/h1-5H,(H,15,16,17)
InChI key   RZQQQWUVWWVDCN-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?