• USA Home
  • CDS014527 - 3-(3-[(tert-Butoxycarbonyl)amino]phenyl)propanoic acid

EMAIL THIS PAGE TO A FRIEND
CDS014527 Sigma-Aldrich

3-(3-[(tert-Butoxycarbonyl)amino]phenyl)propanoic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C14H19NO4

  • Molecular Weight 265.30

  •  MDL number MFCD04037903

  •  PubChem Substance ID 329789013

Purchase

Properties

Related Categories Amines, Building Blocks, C14, Chemical Synthesis, Ethers,
SMILES string   CC(C)(C)OC(=O)Nc1cccc(CCC(O)=O)c1
InChI   1S/C14H19NO4/c1-14(2,3)19-13(18)15-11-6-4-5-10(9-11)7-8-12(16)17/h4-6,9H,7-8H2,1-3H3,(H,15,18)(H,16,17)
InChI key   JZVULXRCJQKJOJ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?