• USA Home
  • CDS014822 - 1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
CDS014822 Sigma-Aldrich

1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C10H12ClN3O2

  • Molecular Weight 241.67

  •  MDL number MFCD00664523

  •  PubChem Substance ID 329789219

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
SMILES string   OC(=O)C1CCN(CC1)c2ccc(Cl)nn2
InChI   1S/C10H12ClN3O2/c11-8-1-2-9(13-12-8)14-5-3-7(4-6-14)10(15)16/h1-2,7H,3-6H2,(H,15,16)
InChI key   DLQSEGIGDCETIS-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?