• USA Home
  • CDS015175 - 1-(4-isobutylphenyl)-3-(3-nitrophenyl)prop-2-en-1-one

EMAIL THIS PAGE TO A FRIEND
CDS015175 Sigma-Aldrich

1-(4-isobutylphenyl)-3-(3-nitrophenyl)prop-2-en-1-one

AldrichCPR

  • Empirical Formula (Hill Notation) C19H19NO3

  • Molecular Weight 309.36

  •  MDL number MFCD00068086

  •  PubChem Substance ID 329789566

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds - Organic Building Blocks, Nitroso Compounds,
SMILES string   CC(C)Cc1ccc(cc1)C(=O)\C=C\c2cccc(c2)[N+]([O-])=O
InChI   1S/C19H19NO3/c1-14(2)12-16-6-9-17(10-7-16)19(21)11-8-15-4-3-5-18(13-15)20(22)23/h3-11,13-14H,12H2,1-2H3/b11-8+
InChI key   FCKXLIVUBCYENE-DHZHZOJOSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?