• USA Home
  • CDS015194 - 4-(4-methoxycarbonylbenzyl)piperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
CDS015194 Sigma-Aldrich

4-(4-methoxycarbonylbenzyl)piperidine hydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C14H20ClNO2

  • Molecular Weight 269.77

  •  MDL number MFCD02179177

  •  PubChem Substance ID 329789584

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
SMILES string   Cl.COC(=O)c1ccc(CC2CCNCC2)cc1
InChI   1S/C14H19NO2.ClH/c1-17-14(16)13-4-2-11(3-5-13)10-12-6-8-15-9-7-12;/h2-5,12,15H,6-10H2,1H3;1H
InChI key   IODVBAPWGFQCFX-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?