• USA Home
  • CDS015919 - 3-benzyloxy-4-fluorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
CDS015919 Sigma-Aldrich

3-benzyloxy-4-fluorobenzaldehyde

AldrichCPR

  • Empirical Formula (Hill Notation) C14H11FO2

  • Molecular Weight 230.23

  •  MDL number MFCD11110161

  •  PubChem Substance ID 329790194

Purchase

Properties

Related Categories Aryl Fluorinated Building Blocks, Building Blocks, C13-C26, C14, Chemical Synthesis,
SMILES string   Fc1ccc(C=O)cc1OCc2ccccc2
InChI   1S/C14H11FO2/c15-13-7-6-12(9-16)8-14(13)17-10-11-4-2-1-3-5-11/h1-9H,10H2
InChI key   SJTDGEBYNOHSTC-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?