• USA Home
  • CDS016327 - 6-(4,6-dioxo-1,4,5,6-tetrahydro-pyrimidin-2-ylamino)-hexanoic acid

EMAIL THIS PAGE TO A FRIEND
CDS016327 Sigma-Aldrich

6-(4,6-dioxo-1,4,5,6-tetrahydro-pyrimidin-2-ylamino)-hexanoic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C10H15N3O4

  • Molecular Weight 241.24

  •  MDL number MFCD00602648

  •  PubChem Substance ID 329790519

Purchase

Properties

Related Categories Building Blocks, C9 to C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
SMILES string   OC(=O)CCCCCNC1=NC(=O)CC(=O)N1
InChI   1S/C10H15N3O4/c14-7-6-8(15)13-10(12-7)11-5-3-1-2-4-9(16)17/h1-6H2,(H,16,17)(H2,11,12,13,14,15)
InChI key   ZAWXEWBTOSOFEQ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?