• USA Home
  • CDS016713 - 1-[1-(3,4-dimethyl-phenyl)-ethyl]-piperazine

EMAIL THIS PAGE TO A FRIEND
CDS016713 Sigma-Aldrich

1-[1-(3,4-dimethyl-phenyl)-ethyl]-piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H22N2

  • Molecular Weight 218.34

  •  MDL number MFCD03724307

  •  PubChem Substance ID 329789891

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
SMILES string   CC(N1CCNCC1)c2ccc(C)c(C)c2
InChI   1S/C14H22N2/c1-11-4-5-14(10-12(11)2)13(3)16-8-6-15-7-9-16/h4-5,10,13,15H,6-9H2,1-3H3
InChI key   KIZHKDIPORTYKA-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?