• USA Home
  • CDS017232 - 1-methyl-4-[4-(1H-pyrazol-5-yl)phenyl]piperazine

EMAIL THIS PAGE TO A FRIEND
CDS017232 Sigma-Aldrich

1-methyl-4-[4-(1H-pyrazol-5-yl)phenyl]piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C14H18N4

  • Molecular Weight 242.32

  •  MDL number MFCD00138784

  •  PubChem Substance ID 329789903

Purchase

Properties

Related Categories Building Blocks, C12 to C37, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks,
SMILES string   CN1CCN(CC1)c2ccc(cc2)-c3ccn[nH]3
InChI   1S/C14H18N4/c1-17-8-10-18(11-9-17)13-4-2-12(3-5-13)14-6-7-15-16-14/h2-7H,8-11H2,1H3,(H,15,16)
InChI key   QIUCNWDIHQUMBA-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?