• USA Home
  • CDS018430 - 2-(2-fluoro-phenyl)-piperazine

EMAIL THIS PAGE TO A FRIEND
CDS018430 Sigma-Aldrich

2-(2-fluoro-phenyl)-piperazine

AldrichCPR

  • Empirical Formula (Hill Notation) C10H13FN2

  • Molecular Weight 180.22

  •  MDL number MFCD07373075

  •  PubChem Substance ID 329792024

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Fluorinated Building Blocks, Halogenated Heterocycles,
SMILES string   Fc1ccccc1C2CNCCN2
InChI   1S/C10H13FN2/c11-9-4-2-1-3-8(9)10-7-12-5-6-13-10/h1-4,10,12-13H,5-7H2
InChI key   JMLSZQAVDKFFCE-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?