• USA Home
  • CDS018506 - (R,S)-N-Fmoc-N′-Boc-imidazolidine-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
CDS018506 Sigma-Aldrich

(R,S)-N-Fmoc-N′-Boc-imidazolidine-2-carboxylic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C24H26N2O6

  • Molecular Weight 438.47

  •  MDL number MFCD01321020

  •  PubChem Substance ID 329791250

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Imidazolines/Imidazolidines
functional group   Boc
  Fmoc
SMILES string   CC(C)(C)OC(=O)N1CCN(C1C(O)=O)C(=O)OCC2c3ccccc3-c4ccccc24
InChI   1S/C24H26N2O6/c1-24(2,3)32-23(30)26-13-12-25(20(26)21(27)28)22(29)31-14-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,19-20H,12-14H2,1-3H3,(H,27,28)
InChI key   IHRMIKPTDYFJCL-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?