• USA Home
  • CDS018601 - 1-(4-fluorophenyl)-4-(4-methylpiperazino)-1-butanone

EMAIL THIS PAGE TO A FRIEND
CDS018601 Sigma-Aldrich

1-(4-fluorophenyl)-4-(4-methylpiperazino)-1-butanone

AldrichCPR

  • Empirical Formula (Hill Notation) C15H21FN2O

  • Molecular Weight 264.34

  •  MDL number MFCD00826065

  •  PubChem Substance ID 329792132

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Fluorinated Building Blocks, Halogenated Heterocycles,
SMILES string   CN1CCN(CCCC(=O)c2ccc(F)cc2)CC1
InChI   1S/C15H21FN2O/c1-17-9-11-18(12-10-17)8-2-3-15(19)13-4-6-14(16)7-5-13/h4-7H,2-3,8-12H2,1H3
InChI key   MZWZUSDMMNOMQQ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?