• USA Home
  • CDS018902 - 2-(4,6-dichloro-2-methyl-1H-indol-3-yl)ethanamine

EMAIL THIS PAGE TO A FRIEND
CDS018902 Sigma-Aldrich

2-(4,6-dichloro-2-methyl-1H-indol-3-yl)ethanamine

AldrichCPR

  • Empirical Formula (Hill Notation) C11H12Cl2N2

  • Molecular Weight 243.13

  •  MDL number MFCD02656500

  •  PubChem Substance ID 329792321

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
SMILES string   CC1=C(CCN)C2=C(Cl)C=C(Cl)C=C2N1
InChI   1S/C11H12Cl2N2/c1-6-8(2-3-14)11-9(13)4-7(12)5-10(11)15-6/h4-5,15H,2-3,14H2,1H3
InChI key   JCTJISIFGZHOFY-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?