• USA Home
  • CDS019289 - 4-(Piperazine-1-carbonyl)-piperidine-1-carboxylic acid tert-butyl ester

EMAIL THIS PAGE TO A FRIEND
CDS019289 Sigma-Aldrich

4-(Piperazine-1-carbonyl)-piperidine-1-carboxylic acid tert-butyl ester

AldrichCPR

  • Empirical Formula (Hill Notation) C15H27N3O3

  • Molecular Weight 297.39

  •  MDL number MFCD07367784

  •  PubChem Substance ID 329792630

Purchase

Properties

Related Categories Building Blocks, C14 to C15, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines,
SMILES string   CC(C)(C)OC(=O)N1CCC(CC1)C(=O)N2CCNCC2
InChI   1S/C15H27N3O3/c1-15(2,3)21-14(20)18-8-4-12(5-9-18)13(19)17-10-6-16-7-11-17/h12,16H,4-11H2,1-3H3
InChI key   YAFVLOOBEOCICL-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?