• USA Home
  • CDS019460 - [4-(2-Amino-phenyl)-piperazin-1-yl]-furan-2-yl-methanone

EMAIL THIS PAGE TO A FRIEND
CDS019460 Sigma-Aldrich

[4-(2-Amino-phenyl)-piperazin-1-yl]-furan-2-yl-methanone

AldrichCPR

  • Empirical Formula (Hill Notation) C15H17N3O2

  • Molecular Weight 271.31

  •  MDL number MFCD08898943

  •  PubChem Substance ID 329792826

Purchase

Properties

Related Categories Building Blocks, C12 to C20+, C14 to C15, Chemical Synthesis, Furans,
SMILES string   Nc1ccccc1N2CCN(CC2)C(=O)c3ccco3
InChI   1S/C15H17N3O2/c16-12-4-1-2-5-13(12)17-7-9-18(10-8-17)15(19)14-6-3-11-20-14/h1-6,11H,7-10,16H2
InChI key   KBQPTOIUYXVHIQ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?