• USA Home
  • CDS020463 - (R,R)-[2-(4′-i-Propyloxazolin-2′-yl)ferrocenyl]diphenylphosphine

EMAIL THIS PAGE TO A FRIEND
CDS020463 Sigma-Aldrich

(R,R)-[2-(4′-i-Propyloxazolin-2′-yl)ferrocenyl]diphenylphosphine

AldrichCPR

  • Empirical Formula (Hill Notation) C28H28FeNOP

  • Molecular Weight 481.35

  •  MDL number MFCD17018796

  •  PubChem Substance ID 329793793

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Oxazolines/Oxazolidines
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.CC(C)[C@@H]2COC(=N2)[C]3[CH][CH][CH][C]3P(c4ccccc4)c5ccccc5
InChI   1S/C23H23NOP.C5H5.Fe/c1-17(2)21-16-25-23(24-21)20-14-9-15-22(20)26(18-10-5-3-6-11-18)19-12-7-4-8-13-19;1-2-4-5-3-1;/h3-15,17,21H,16H2,1-2H3;1-5H;/t21-;;/m0../s1
InChI key   FELCHLOAEMNOQM-FGJQBABTSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?