• USA Home
  • CDS021599 - Triciribine phosphate

EMAIL THIS PAGE TO A FRIEND
CDS021599 Sigma-Aldrich

Triciribine phosphate

  • Empirical Formula (Hill Notation) C13H17N6O7P

  • Molecular Weight 400.28

  •  MDL number MFCD00866447

  •  PubChem Substance ID 329794862

Purchase

Properties

description   AldrichCPR
SMILES string   NC1=NN(C)C2=NC=NC3=C2C1=CN3[C@@H]([C@H](O)[C@@H]4O)O[C@@H]4COP(O)(O)=O
InChI   1S/C13H17N6O7P/c1-18-11-7-5(10(14)17-18)2-19(12(7)16-4-15-11)13-9(21)8(20)6(26-13)3-25-27(22,23)24/h2,4,6,8-9,13,20-21H,3H2,1H3,(H2,14,17)(H2,22,23,24)/t6-,8-,9-,13-/m1/s1
InChI key   URLYINUFLXOMHP-HTVVRFAVSA-N
Gene Information   human ... AKT1(207), AKT2(208), AKT3(10000)

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?