• USA Home
  • CLS3895BC - Corning® 1536 well plate, with generic barcode

EMAIL THIS PAGE TO A FRIEND
CLS3895BC Sigma-Aldrich

Corning® 1536 well plate, with generic barcode

size 1536 wells, NBS surface, clear polystyrene wells flat bottom, non-sterile

Synonym: 1536 multiwell plate, 1536 well barcode plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate with generic barcode, 1536 well plate

Purchase

Properties

Related Categories 1536 Well Plates, Corning, Corning 1536-Well Microplates, Corning 1536-Well Multiwell Plates, Corning Microplates,
material   black plate
  clear polystyrene wells flat bottom
  polystyrene plate
sterility   non-sterile
feature   lid: no
packaging   pack of 10
  pkg of 10 ea
  case of 50 ea
  case of 50
mfr. no.   Corning, 3895BC
maximum volume   12.5 μL
size   1536 wells
working volume   8 μL
color   clear
matrix active group   NBS surface surface treatment
  non-treated surface treatment

Description

General description

Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

• Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
• Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
• Bar Code labeling specifications
• 2.75" x 0.25" label size
• Code 128
• Code 3 of 9
• 10 mil narrow bar element (x-dimension = 0.010")

Legal Information

Corning is a registered trademark of Corning, Inc.

NBS is a trademark of Corning, Inc.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?