• USA Home
  • D114 - (±)-1-(2,5-Dimethoxyphenyl)-2-aminopropane hydrochloride

EMAIL THIS PAGE TO A FRIEND
D114 Sigma-Aldrich

(±)-1-(2,5-Dimethoxyphenyl)-2-aminopropane hydrochloride

Synonym: DMA

Purchase

Properties

drug control   USDEA Schedule I; Home Office Schedule 1; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
Gene Information   human ... SLC9A1(6548)
rat ... Slc9a1(24782), Slc9a2(24783), Slc9a3(24784)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?