• USA Home
  • D11809 - 1,5-Diaminoanthraquinone

EMAIL THIS PAGE TO A FRIEND
D11809 Aldrich

1,5-Diaminoanthraquinone

97%

Purchase

Properties

InChI Key   VWBVCOPVKXNMMZ-UHFFFAOYSA-N
assay   97%
mp   >300 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
CB6400000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?