• USA Home
  • D19206 - 2,4-Diamino-6-hydroxypyrimidine

EMAIL THIS PAGE TO A FRIEND
D19206 Sigma-Aldrich

2,4-Diamino-6-hydroxypyrimidine

96%

Synonym: 2,4-Diamino-6-hydroxypyrimidine, 2,6-Diamino-4-pyrimidinol, 2,6-Diamino-4-pyrimidinone

  • CAS Number 56-06-4

  • Empirical Formula (Hill Notation) C4H6N4O

  • Molecular Weight 126.12

  •  Beilstein/REAXYS Number 125006

  •  EC Number 200-254-4

  •  MDL number MFCD00006098

  •  PubChem Substance ID 24278824

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
Quality Level   200
assay   96%
mp   285-286 °C (dec.) (lit.)
storage temp.   room temp
SMILES string   Nc1cc(O)nc(N)n1
InChI   1S/C4H6N4O/c5-2-1-3(9)8-4(6)7-2/h1H,(H5,5,6,7,8,9)
InChI key   SWELIMKTDYHAOY-UHFFFAOYSA-N

Description

Application

2,4-Diamino-6-hydroxypyrimidine can be used as a reactant:
• To prepare biologically significant phosphonomethoxy pyrimidine derivatives.
• To synthesize pharmacologically important pyrrolo[2,3-d]pyrimidine derivatives.
• In the total synthesis of a natural product Q base or queuine, which is commonly found in tRNA of plants and animals.

Packaging

25, 100 g in poly bottle

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. D19206.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?