• USA Home
  • D60850 - 2,4-Dichloro-3,5-dinitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
D60850 Sigma-Aldrich

2,4-Dichloro-3,5-dinitrobenzoic acid

95%

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
assay   95%
mp   212-213 °C (lit.)
SMILES string   OC(=O)c1cc(c(Cl)c(c1Cl)[N+]([O-])=O)[N+]([O-])=O
InChI   1S/C7H2Cl2N2O6/c8-4-2(7(12)13)1-3(10(14)15)5(9)6(4)11(16)17/h1H,(H,12,13)
InChI key   OCJYCLPVZHBZRL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?