EMAIL THIS PAGE TO A FRIEND
E35257 Sigma-Aldrich

Thimerosal

97%

Synonym: 2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

  • CAS Number 54-64-8

  • Linear Formula 2-(C2H5HgS)C6H4CO2Na

  • Molecular Weight 404.81

  •  Beilstein/REAXYS Number 8169555

  •  EC Number 200-210-4

  •  MDL number MFCD00013062

Purchase

Properties

assay   97%
mp   234-237 °C (dec.) (lit.)
SMILES string   [Na+].CC[Hg]Sc1ccccc1C([O-])=O
InChI   1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2
InChI key   RTKIYNMVFMVABJ-UHFFFAOYSA-L

Description

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2025 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
482 °F
Flash Point(C) 
250 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T5125 Thimerosal, 97.0-101.0% (on dried substance, titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?