• USA Home
  • G11705 - Guanidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
G11705 Sigma-Aldrich

Guanidine hydrochloride

92%

Synonym: Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

  • CAS Number 50-01-1

  • Linear Formula NH2C(=NH)NH2 · HCl

  • Molecular Weight 95.53

  •  Beilstein/REAXYS Number 3591990

  •  EC Number 200-002-3

  •  MDL number MFCD00013026

  •  PubChem Substance ID 329799831

Purchase

Properties

assay   92%
mp   180-185 °C (lit.)
density   1.3 g/cm3 (lit.)
SMILES string   Cl[H].NC(N)=N
InChI   1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H
InChI key   PJJJBBJSCAKJQF-UHFFFAOYSA-N
Gene Information   human ... KCNA1(3736), KCNA10(3744), KCNA2(3737), KCNA3(3738), KCNA4(3739), KCNA5(3741), KCNA6(3742), KCNA7(3743), KCNB1(3745), KCNB2(9312), KCNC1(3746), KCNC2(3747), KCNC3(3748), KCNC4(3749), KCND1(3750), KCND2(3751), KCND3(3752), KCNF1(3754), KCNG1(3755), KCNG2(26251), KCNG3(170850), KCNG4(93107), KCNH1(3756), KCNH2(3757), KCNH3(23416), KCNH4(23415), KCNH5(27133), KCNH6(81033), KCNH7(90134), KCNH8(131096), KCNQ1(3784), KCNQ2(3785), KCNQ3(3786), KCNQ4(9132), KCNQ5(56479), KCNS1(3787), KCNS2(3788), KCNS3(3790), KCNV1(27012), KCNV2(169522)

Description

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MF4300000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G4505 Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?