• USA Home
  • GF00902225 - Titanium/Aluminum/Vanadium

EMAIL THIS PAGE TO A FRIEND
GF00902225

Titanium/Aluminum/Vanadium

tube, Ti 94.5%/Al 3%/V 2.5%, outside diameter 6.35 mm, length 200 mm, wall thickness 0.71 mm

Synonym: TI047200

  •  NACRES NA.23

Purchase

Properties

form   tubes
mfr. no.   Goodfellow, 009-022-25
L   200 mm
O.D.   6.35 mm
wall thickness   0.71 mm
storage temp.   room temp

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?