• USA Home
  • GF06141170 - Polymethylpentene

EMAIL THIS PAGE TO A FRIEND
GF06141170

Polymethylpentene

TPX®, rod, diam. 12.7 mm, L 400 mm, natural

Synonym: ME317916

  •  NACRES NA.23

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 061-411-70
L   400 mm
diam.   12.7 mm
color   natural
storage temp.   room temp

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

TPX is a registered trademark of Mitsui Petrochemical Industries, Ltd.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?