EMAIL THIS PAGE TO A FRIEND
GF07601059

Praseodymium

wire reel, 50mm, diameter 1.0mm, hard, 99.9%

Synonym: PR005110, Praseodymium

  • CAS Number 7440-10-0

  • Empirical Formula (Hill Notation) Pr

  • Molecular Weight 140.91

  •  MDL number MFCD00011174

  •  PubChem Substance ID 329801201

  •  NACRES NA.23

Purchase

Properties

assay   99.9%
form   wire
mfr. no.   Goodfellow, 076-010-59
resistivity   68 μΩ-cm, 20°C
size L × diam.   50 mm × 1.0 mm
bp   3520 °C (lit.)
mp   931 °C (lit.)
density   6.71 g/mL at 25 °C (lit.)
SMILES string   [Pr]
InChI   1S/Pr
InChI key   PUDIUYLPXJFUGB-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

RIDADR 
UN1383- class 4.2 - PG 1- Pyrophoric metal, n-o.s., HI: all
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?