• USA Home
  • GF08412768 - Copper/Beryllium

EMAIL THIS PAGE TO A FRIEND
GF08412768

Copper/Beryllium

foil, Cu 98%/Be 2%, thickness 0.015 mm, 8 mm diameter, temper as rolled

Synonym: CU070271

  •  NACRES NA.23

Purchase

Properties

form   foil
mfr. no.   Goodfellow, 084-127-68
diam.   8 mm
thickness   0.015 mm
diameter   8 mm
storage temp.   room temp

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?