EMAIL THIS PAGE TO A FRIEND
GF18711693

Silver

insulated wire, 2m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%

Synonym: AG005860, Silver

Purchase

Properties

assay   99.99%
form   wire
mfr. no.   Goodfellow, 187-116-93
resistivity   1.59 μΩ-cm, 20°C
L × diam. × thickness   2 m × 0.25 mm × 0.015 mm
bp   2212 °C (lit.)
mp   960 °C (lit.)
density   10.49 g/cm3 (lit.)
SMILES string   [Ag]
InChI   1S/Ag
InChI key   BQCADISMDOOEFD-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?