EMAIL THIS PAGE TO A FRIEND
GF22038724

Polypropylene

(PP), film, thickness 0.04 mm, L 1 m, condition biaxially oriented

Synonym: PP301341

  •  NACRES NA.23

Purchase

Properties

form   film
mfr. no.   Goodfellow, 220-387-24
L   1 m
thickness   0.04 mm
storage temp.   room temp

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?