EMAIL THIS PAGE TO A FRIEND
GF25529407 Aldrich

Samarium

foil, 50x50mm, thickness 0.1mm, as rolled, 99%

Synonym: SM000210, Samarium

  • CAS Number 7440-19-9

  • Empirical Formula (Hill Notation) Sm

  • Molecular Weight 150.36

  •  MDL number MFCD00011233

  •  PubChem Substance ID 329803928

  •  NACRES NA.23

Purchase

Properties

InChI Key   KZUNJOHGWZRPMI-UHFFFAOYSA-N
assay   99%
form   foil
mfr. no.   Goodfellow, 255-294-07
resistivity   91.4 μΩ-cm, 0°C
size size × thickness   50x50 mm × 0.1 mm
bp   1794 °C (lit.)
mp   1074 °C (lit.)
density   7.47 g/mL at 25 °C (lit.)

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?