EMAIL THIS PAGE TO A FRIEND
GF31780571

Carbon

microleaf, 24x70mm, thinness 5.0μm, specific density 1000μg/cm2, 99.997%

Synonym: Activated charcoal, C 004995, Charcoal activated

  • CAS Number 7440-44-0

  • Empirical Formula (Hill Notation) C

  • Molecular Weight 12.01

  •  MDL number MFCD00133992

  •  PubChem Substance ID 329804836

  •  NACRES NA.23

Purchase

Properties

vapor pressure   <0.1 mmHg ( 20 °C)
assay   99.997%
form   foil (microleaf)
autoignition temp.   842 °F
mfr. no.   Goodfellow, 317-805-71
resistivity   1375 μΩ-cm, 20°C (graphite)
size L × W × thickness   24 × 70 mm (5.0)  μm
mp   3550 °C (lit.)
SMILES string   [C]
InChI   1S/C
InChI key   OKTJSMMVPCPJKN-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?