EMAIL THIS PAGE TO A FRIEND
GF35817217

Aluminum

sphere, 200pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%

Synonym: AL006830, Aluminum

  • CAS Number 7429-90-5

  • Empirical Formula (Hill Notation) Al

  • Molecular Weight 26.98

  •  MDL number MFCD00134029

  •  PubChem Substance ID 329805448

  •  NACRES NA.23

Purchase

Properties

assay   95.2%
form   spheres
autoignition temp.   1400 °F
mfr. no.   Goodfellow, 358-172-17
resistivity   2.6548 μΩ-cm
diameter   3.0 mm
bp   2460 °C (lit.)
mp   660.37 °C (lit.)
density   2.7 g/mL at 25 °C (lit.)
SMILES string   [Al]
InChI   1S/Al
InChI key   XAGFODPZIPBFFR-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?