• USA Home
  • GF37017402 - Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

EMAIL THIS PAGE TO A FRIEND
GF37017402

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

245mm tube, 7.5mm outside diameter, 0.7mm wall thickness, 6.1mm inside diameter, annealed

Synonym: NI027400

  •  NACRES NA.23

Purchase

Properties

form   tubes
mfr. no.   Goodfellow, 370-174-02
size L × wall thickness   245 mm × 0.7 mm
size O.D. × I.D.   7.5 mm × 6.1 mm

Description

General description

For updated SDS information please visit www.goodfellow.com.

Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation

Inconel is a registered trademark of Special Metals Corporation

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?