• USA Home
  • GF58672200 - Stainless Steel - AISI 302 alloy, FeCr18Ni8

EMAIL THIS PAGE TO A FRIEND
GF58672200

Stainless Steel - AISI 302 alloy, FeCr18Ni8

100m wire, 1.0mm diameter, hard

Synonym: FE205150

  •  NACRES NA.23

Purchase

Properties

form   wire
mfr. no.   Goodfellow, 586-722-00
L × diam.   100 m × 1.0 mm

Description

General description

For updated SDS information please visit www.goodfellow.com.

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?