EMAIL THIS PAGE TO A FRIEND
GF72504716

Carbon

lump, 1.5 mm max. lump size, weight 100 g, purity 99.8%

Synonym: Activated charcoal, C 006100, Charcoal activated

  • CAS Number 7440-44-0

  • Empirical Formula (Hill Notation) C

  • Molecular Weight 12.01

  •  MDL number MFCD00133992

  •  PubChem Substance ID 329810865

  •  NACRES NA.23

Purchase

Properties

vapor pressure   <0.1 mmHg ( 20 °C)
assay   99.8%
form   lumps
autoignition temp.   842 °F
mfr. no.   Goodfellow, 725-047-16
resistivity   1375 μΩ-cm, 20°C (graphite)
weight   100 g
max. lump size   1.5 mm
mp   3550 °C (lit.)
SMILES string   [C]
InChI   1S/C
InChI key   OKTJSMMVPCPJKN-UHFFFAOYSA-N

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?